siRNA Details
Gene Symbol: Wnt5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100489.1 | NP_001093959.1 | 4 | 544 | ||
XM_006237179.1 | XP_006237241.1 | 3 | 422 | |||
GenBank mRNA | AF481944.1 | - | 2 | 69 | ||
BC161860.1 | - | 687 | 7096336 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
