siRNA Details
Gene Symbol: Pxmp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172223.2 | NP_757377.1 | 3 | 288 | ||
GenBank mRNA | AY151834.1 | - | 3 | 266 | ||
BC081699.1 | - | 3 | 288 | 7193564 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
