siRNA Details
Gene Symbol: Impa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172224.1 | NP_757378.1 | 2 | 347 | ||
XM_006254856.2 | XP_006254918.1 | 2 | 3666 | |||
GenBank mRNA | AY160191.1 | - | 2 | 347 | ||
BC083544.1 | - | 2 | 321 | 7191219 |
