siRNA Details
Gene Symbol: Afm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172320.1 | NP_758823.1 | 12 | 1631 | ||
XM_008770001.1 | XP_008768223.1 | 12 | 1679 | |||
GenBank mRNA | BC091391.1 | - | 9 | 1196 | ||
FQ213773.1 | - | 8 | 1067 | |||
FQ219466.1 | - | 9 | 1165 |
