siRNA Details
Gene Symbol: Zfp161 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172325.2 | NP_758828.2 | 373 | |||
GenBank mRNA | AY157624.1 | - | 1 | 292 |

More Information
Other Species Targeted

Gene Ontology Categories:
