siRNA Details
Gene Symbol: Tac4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172328.2 | NP_758831.1 | 2 | 149 | ||
GenBank mRNA | AF521561.1 | - | 2 | 149 | ||
AY471575.1 | - | 2 | 149 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
