siRNA Details
Gene Symbol: Chmp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172331.1 | NP_758834.1 | 1, 2 | 71 | ||
GenBank mRNA | AY150169.1 | - | 1, 2 | 71 | ||
FQ215758.1 | - | 1, 2 | 85 | |||
FQ219218.1 | - | 1, 2 | 90 | |||
FQ234202.1 | - | 1, 2 | 148 |

More Information
Other Species Targeted

Gene Ontology Categories:
