siRNA Details
Gene Symbol: Atf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172336.3 | NP_758839.3 | 2 | 797 | ||
XM_006228980.1 | XP_006229042.1 | 3 | 1075 | |||
GenBank mRNA | AY123225.1 | - | 2 | 797 | ||
BC061786.1 | - | 3 | 1075 | 5623784 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
