siRNA Details
Gene Symbol: Sorbs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005762.1 | NP_001005762.1 | 2 | 192 | ||
XM_006252241.2 | XP_006252303.1 | 3 | 1876 | |||
XM_006252242.1 | XP_006252304.1 | 2 | 283 | |||
XM_006252246.1 | XP_006252308.1 | 2 | 283 | |||
GenBank mRNA | BC083666.1 | - | 2 | 192 | 7135547 |
