siRNA Details
Gene Symbol: Rnf34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004075.1 | NP_001004075.1 | 2 | 269 | ||
XM_006249360.2 | XP_006249422.1 | 2 | 802 | |||
XM_006249361.2 | XP_006249423.1 | 3 | 479 | |||
XM_008769214.1 | XP_008767436.1 | 2 | 187 | |||
GenBank mRNA | AY157968.1 | - | 1 | 174 | ||
BC079044.1 | - | 2 | 269 | 7115027 |
