siRNA Details
Gene Symbol: Aqp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173105.1 | NP_775128.1 | 1 | 688 | ||
GenBank mRNA | AB023644.1 | - | 1 | 688 | ||
BC078694.1 | - | 1 | 715 | 7111950 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Aqp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173105.1 | NP_775128.1 | 1 | 688 | ||
GenBank mRNA | AB023644.1 | - | 1 | 688 | ||
BC078694.1 | - | 1 | 715 | 7111950 |