siRNA Details
Gene Symbol: Wfdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173109.1 | NP_775132.1 | 3 | 382 | ||
XM_008762473.1 | XP_008760695.1 | 3 | 387 | |||
GenBank mRNA | AJ515239.1 | - | 3 | 382 | ||
AJ515241.1 | - | 2 | 202 | |||
FQ222047.1 | - | 1 | 140 |
