siRNA Details
Gene Symbol: Parm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173114.1 | NP_775137.1 | 2 | 261 | ||
GenBank mRNA | AJ010750.1 | - | 2 | 283 | ||
BC060539.1 | - | 2 | 287 | 6890269 | ||
FQ219991.1 | - | 2 | 284 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
