siRNA Details
Gene Symbol: Npc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173118.2 | NP_775141.2 | 3, 4 | 463 | ||
GenBank mRNA | AJ515237.1 | - | 3, 4 | 398 | ||
BC058132.1 | - | 3, 4 | 426 | 6920130 | ||
FQ214040.1 | - | 3, 4 | 384 | |||
FQ216545.1 | - | 3, 4 | 388 | |||
FQ219301.1 | - | 3, 4 | 405 | |||
FQ219863.1 | - | 3, 4 | 396 | |||
FQ220660.1 | - | 3, 4 | 384 | |||
FQ220919.1 | - | 3, 4 | 390 | |||
FQ228456.1 | - | 3, 4 | 388 | |||
FQ229422.1 | - | 3, 4 | 443 | |||
FQ229782.1 | - | 3, 4 | 384 | |||
FQ229783.1 | - | 3, 4 | 386 |
