siRNA Details
Gene Symbol: Hsbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173119.2 | NP_775142.1 | 2 | 179 | ||
GenBank mRNA | AF522937.1 | - | 2 | 126 | ||
BC058131.1 | - | 2 | 164 | 6919748 | ||
EX491168.1 | - | 2 | 179 | |||
FQ211891.1 | - | 2 | 147 | |||
FQ212621.1 | - | 2 | 151 | |||
FQ213728.1 | - | 2 | 121 | |||
FQ216175.1 | - | 2 | 158 | |||
FQ229155.1 | - | 2 | 164 | |||
FQ229754.1 | - | 2 | 138 | |||
FQ230047.1 | - | 2 | 136 | |||
FQ230722.1 | - | 2 | 138 | |||
FQ233773.1 | - | 2 | 137 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
