siRNA Details
Gene Symbol: Vimp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173120.2 | NP_775143.2 | 3 | 317 | ||
GenBank mRNA | AF367467.1 | - | 3 | 327 | ||
BC059111.1 | - | 3 | 321 | 6920258 | ||
FQ210095.1 | - | 3 | 309 | |||
FQ218853.1 | - | 3 | 327 | |||
FQ219944.1 | - | 3 | 309 | |||
FQ220082.1 | - | 3 | 312 | |||
FQ225575.1 | - | 3 | 327 | |||
FQ229642.1 | - | 3 | 309 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
