siRNA Details
Gene Symbol: Cyp4f5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173124.1 | NP_775147.1 | 474 | |||
XM_006241051.2 | XP_006241113.1 | 4, 5 | 543 | |||
XM_008765158.1 | XP_008763380.1 | 4, 5 | 545 | |||
GenBank mRNA | BC129085.1 | - | 4, 5 | 435 | 7375658 | |
U39207.1 | - | 474 |
Gene Symbol: Cyp4f37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271352.1 | NP_001258281.1 | 5, 6 | 794 |
Gene Symbol: LOC100912471 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006241097.2 | XP_006241159.1 | 2, 3 | 110 |
Gene Symbol: LOC691320 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006241045.1 | XP_006241107.1 | 1, 2 | 77 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
