siRNA Details
Gene Symbol: Smim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173126.3 | NP_775149.1 | 2 | 1431 | ||
XM_006254762.2 | XP_006254824.1 | 4 | 2114 | |||
XM_006254763.2 | XP_006254825.1 | 4 | 2117 | |||
XM_006254765.2 | XP_006254827.1 | 4 | 1643 | |||
XM_006254767.1 | XP_006254829.1 | 2 | 1117 | |||
XM_006254769.2 | XP_006254831.1 | 2 | 1320 | |||
XM_008772096.1 | XP_008770318.1 | 3 | 2245 | |||
XM_008772097.1 | XP_008770319.1 | 2 | 2053 | |||
GenBank mRNA | AF313411.1 | - | 2 | 1229 | ||
BC070515.1 | - | 2 | 1430 | 7100997 |
