siRNA Details
Gene Symbol: Phax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173133.2 | NP_775156.2 | 2 | 177 | ||
GenBank mRNA | BC086410.1 | - | 2 | 147 | ||
BC097932.1 | - | 2 | 153 | 7386998 | ||
CB706455.1 | - | 2 | 376 | |||
X67877.1 | - | 2 | 177 |
