siRNA Details
Gene Symbol: Np4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173299.2 | NP_775421.1 | 2 | 224 | ||
XM_006253366.1 | XP_006253428.1 | 2 | 264 | |||
GenBank mRNA | U16684.1 | - | 2 | 225 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
