siRNA Details
Gene Symbol: Hsd17b6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173305.1 | NP_775427.1 | 3 | 822 | ||
XM_006240739.2 | XP_006240801.2 | 3 | 618 | |||
GenBank mRNA | BC088104.1 | - | 3 | 772 | 7326228 | |
BC091114.1 | - | 3 | 734 | 7321553 | ||
U89280.1 | - | 3 | 822 |
Gene Symbol: LOC100362350 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769893.1 | XP_008768115.1 | 2 | 604 |
