siRNA Details
Gene Symbol: Zfp455 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173314.2 | NP_775436.2 | 4 | 1192 | ||
XM_006232101.2 | XP_006232163.1 | 4 | 1428 | |||
GenBank mRNA | BC161833.1 | - | 5 | 1024 | ||
U67083.1 | - | 1180 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
