siRNA Details
Gene Symbol: Zfand4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173332.2 | NP_775454.2 | 9 | 2085 | ||
XM_006237141.2 | XP_006237203.1 | 10 | 2608 | |||
XM_006237142.2 | XP_006237204.1 | 9 | 3664 | |||
XM_006237143.2 | XP_006237205.1 | 9 | 2462 | |||
XM_006237145.2 | XP_006237207.1 | 10 | 2947 | |||
XM_006237146.1 | XP_006237208.1 | 7 | 1766 | |||
XM_008763224.1 | XP_008761446.1 | 10 | 2926 | |||
XM_008763225.1 | XP_008761447.1 | 11 | 2679 | |||
XM_008763226.1 | XP_008761448.1 | 9 | 2779 | |||
XM_008763227.1 | XP_008761449.1 | 7 | 1893 | |||
GenBank mRNA | BC162051.1 | - | 9 | 2085 | 7112072 |
