siRNA Details
Gene Symbol: Pkmyt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105766.1 | NP_001099236.1 | 3 | 724 | ||
XM_006245868.2 | XP_006245930.1 | 2 | 377 | |||
GenBank mRNA | BC169008.1 | - | 3 | 630 | 7387434 |

More Information
Other Species Targeted

Gene Ontology Categories:
