siRNA Details
Gene Symbol: Pgp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001169152.1 | NP_001162623.1 | 2 | 952 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_025954, AK013578, AK012010, BC040100, BC083113, and others.
Gene Ontology Categories:
