siRNA Details
Gene Symbol: Pigq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007607.1 | NP_001007608.1 | 4 | 948 | ||
XM_006245942.2 | XP_006246004.1 | 4 | 1027 | |||
XM_008767559.1 | XP_008765781.1 | 4 | 967 | |||
XM_008767560.1 | XP_008765782.1 | 4 | 994 | |||
GenBank mRNA | BC081875.1 | - | 4 | 948 | 7107681 |

More Information
Other Species Targeted

Gene Ontology Categories:
