siRNA Details
Gene Symbol: LOC287167 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013853.1 | NP_001013875.1 | 1 | 49 | ||
GenBank mRNA | FQ211031.1 | - | 1 | 80 | ||
FQ212394.1 | - | 1 | 80 | |||
FQ218263.1 | - | 1 | 80 | |||
FQ224052.1 | - | 1 | 80 | |||
FQ226501.1 | - | 1 | 82 | |||
FQ228302.1 | - | 1 | 80 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
