siRNA Details
Gene Symbol: Rars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105777.2 | NP_001099247.2 | 6, 7 | 715 | ||
GenBank mRNA | BC161929.1 | - | 6, 7 | 701 | 8375237 | |
CF110623.1 | - | 2, 3 | 194 |
