siRNA Details
Gene Symbol: Clk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013041.1 | NP_001013059.1 | 6 | 669 | ||
XM_006246234.2 | XP_006246296.1 | 7 | 879 | |||
XM_006246236.1 | XP_006246298.1 | 10 | 1082 | |||
XM_006246237.1 | XP_006246299.1 | 8 | 784 | |||
XM_006246238.2 | XP_006246300.1 | 7 | 2299 | |||
XM_008767670.1 | XP_008765892.1 | 8 | 1753 | |||
XM_008767671.1 | XP_008765893.1 | 7 | 1675 | |||
XM_008767672.1 | XP_008765894.1 | 7 | 718 | |||
XM_008767673.1 | XP_008765895.1 | 6 | 2371 | |||
XM_008767674.1 | XP_008765896.1 | 5 | 2158 | |||
XM_008767675.1 | XP_008765897.1 | 8 | 945 | |||
XM_008767676.1 | XP_008765898.1 | 6 | 670 | |||
GenBank mRNA | BC079006.1 | - | 6 | 669 | 7113075 | |
FQ223275.1 | - | 9 | 2729 |
