siRNA Details
Gene Symbol: LOC103690013 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006246240.2 | XP_006246302.1 | 3 | 335 |
Gene Symbol: LOC100910210 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | FQ222023.1 | - | 3 | 316 |
Gene Symbol: Nhp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105779.1 | NP_001099249.1 | 4 | 233 | ||
XM_003750818.3 | XP_003750866.1 | 3 | 335 | |||
GenBank mRNA | BC162010.1 | - | 3 | 274 | 9091322 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
