siRNA Details
Gene Symbol: Sh3bp5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127581.1 | NP_001121053.1 | 4 | 966 | ||
XM_008767708.1 | XP_008765930.1 | 5 | 1083 | |||
XM_008767709.1 | XP_008765931.1 | 5 | 1053 | |||
XM_008767710.1 | XP_008765932.1 | 5 | 1091 | |||
GenBank mRNA | BC166447.1 | - | 4 | 966 | 8364461 |

More Information
Other Species Targeted

