siRNA Details
Gene Symbol: Mrpl55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105782.1 | NP_001099252.1 | 4 | 539 | ||
XM_006246446.2 | XP_006246508.1 | 4 | 580 | |||
XM_006246447.2 | XP_006246509.1 | 4 | 570 | |||
XM_008767780.1 | XP_008766002.1 | 4 | 932 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
