siRNA Details
Gene Symbol: Dhrs7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008507.1 | NP_001008507.1 | 2 | 152 | ||
XM_006246472.2 | XP_006246534.1 | 3 | 360 | |||
XM_006246473.2 | XP_006246535.1 | 2 | 1021 | |||
XM_006246474.2 | XP_006246536.1 | 2 | 489 | |||
XM_006246475.2 | XP_006246537.1 | 2 | 265 | |||
XM_008767786.1 | XP_008766008.1 | 2 | 348 | |||
GenBank mRNA | BC086453.1 | - | 2 | 152 | 7305216 | |
EF445633.1 | - | 3 | 264 |
