siRNA Details
Gene Symbol: LOC100911746 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750843.3 | XP_003750891.1 | 3 | 349 | ||
XM_006246547.2 | XP_006246609.1 | 3 | 979 | |||
XM_006246548.2 | XP_006246610.1 | 3 | 985 |
Gene Symbol: B9d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105786.1 | NP_001099256.1 | 3 | 273 | ||
XM_006246553.2 | XP_006246615.1 | 979 | ||||
XM_006246554.2 | XP_006246616.1 | 985 | ||||
GenBank mRNA | BC169130.1 | - | 3 | 231 | 6889289 |
