siRNA Details
Gene Symbol: Aloxe3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105793.1 | NP_001099263.1 | 12 | 1606 | ||
XM_006246617.2 | XP_006246679.1 | 13 | 2238 | |||
XM_006246619.2 | XP_006246681.1 | 12 | 1678 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
