siRNA Details
Gene Symbol: Tnfsf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009623.1 | NP_001009623.1 | 5 | 841 | ||
XM_006246625.2 | XP_006246687.1 | 6 | 1426 | |||
XM_008767796.1 | XP_008766018.1 | 5 | 1582 | |||
XM_008767797.1 | XP_008766019.1 | 5 | 1579 | |||
XM_008767798.1 | XP_008766020.1 | 4 | 1534 | |||
XM_008767799.1 | XP_008766021.1 | 4 | 1531 | |||
XM_008767800.1 | XP_008766022.1 | 5 | 1582 | |||
XM_008767801.1 | XP_008766023.1 | 5 | 1579 | |||
XM_008767802.1 | XP_008766024.1 | 4 | 1534 | |||
GenBank mRNA | BC087135.1 | - | 5 | 841 | 7123934 | |
FQ215356.1 | - | 5 | 611 |

More Information
Other Species Targeted

Gene Ontology Categories:
