siRNA Details
Gene Symbol: Acap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105796.2 | NP_001099266.2 | 17 | 1756 | ||
GenBank mRNA | FQ230675.1 | - | 1 | 13 | ||
FQ231017.1 | - | 1 | 13 | |||
FQ231544.1 | - | 1 | 13 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
