siRNA Details
Gene Symbol: Ctdnep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100494.1 | NP_001093964.1 | 1, 2 | 314 | ||
XM_008767804.1 | XP_008766026.1 | 1, 2 | 343 | |||
GenBank mRNA | BU671232.1 | - | 1, 2 | 314 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
