siRNA Details
Gene Symbol: Zzef1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006246857.2 | XP_006246919.1 | 11 | 1817 | ||
XM_237787.8 | XP_237787.4 | 11 | 1817 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
