siRNA Details
Gene Symbol: Blmh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034163.1 | NP_001029335.1 | 5 | 704 | ||
GenBank mRNA | BC128701.1 | - | 5 | 589 | 7379958 | |
CB772927.1 | - | 3 | 276 | |||
CK363809.1 | - | 5 | 631 | |||
D87336.1 | - | 5 | 607 |
