siRNA Details
Gene Symbol: Vezf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768166.1 | XP_008766388.1 | 2 | 225 | ||
XM_008768167.1 | XP_008766389.1 | 2 | 225 |

More Information
Other Species Targeted

Gene Ontology Categories:
