siRNA Details
Gene Symbol: Samd14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024966.1 | NP_001020137.1 | 8 | 1012 | ||
GenBank mRNA | BC091329.1 | - | 8 | 1012 | 7313977 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
