siRNA Details
Gene Symbol: Spop | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100496.1 | NP_001093966.1 | 8 | 886 | ||
XM_006247180.2 | XP_006247242.1 | 9 | 1690 | |||
XM_006247181.2 | XP_006247243.1 | 10 | 997 | |||
XM_006247182.2 | XP_006247244.1 | 9 | 946 | |||
XM_008767968.1 | XP_008766190.1 | 9 | 1042 | |||
XM_008767969.1 | XP_008766191.1 | 10 | 964 | |||
GenBank mRNA | BC091435.1 | - | 6 | 612 | ||
BC166743.1 | - | 8 | 886 | 7938440 | ||
FQ217100.1 | - | 10 | 1194 |
