siRNA Details
Gene Symbol: Phospho1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105833.1 | NP_001099303.1 | 3 | 745 | ||
XM_006247184.2 | XP_006247246.1 | 3 | 608 | |||
XM_008767970.1 | XP_008766192.1 | 3 | 1178 | |||
GenBank mRNA | BC169002.1 | - | 3 | 688 | 7455281 |

More Information
Other Species Targeted

Gene Ontology Categories:
