siRNA Details
Gene Symbol: Cisd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105835.1 | NP_001099305.1 | 4 | 806 | ||
XM_006247269.2 | XP_006247331.1 | 3 | 703 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
