siRNA Details
Gene Symbol: Pcgf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105836.2 | NP_001099306.1 | 5 | 532 | ||
XM_008767971.1 | XP_008766193.1 | 6 | 491 | |||
XM_008767972.1 | XP_008766194.1 | 6 | 767 | |||
XM_008767973.1 | XP_008766195.1 | 6 | 837 | |||
GenBank mRNA | BC167059.1 | - | 5 | 532 | 7445511 |
