siRNA Details
Gene Symbol: Pcgf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767971.1 | XP_008766193.1 | 10 | 1330 | ||
XM_008767972.1 | XP_008766194.1 | 10 | 1606 | |||
XM_008767973.1 | XP_008766195.1 | 10 | 1676 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001163307, NM_009545, NM_001163308, AK142837.
Gene Ontology Categories:
