siRNA Details
Gene Symbol: Krt15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004022.2 | NP_001004022.1 | 4 | 907 | ||
GenBank mRNA | BC101868.1 | - | 4 | 907 | 7938504 |

More Information
Other Species Targeted

Gene Ontology Categories:
