siRNA Details
Gene Symbol: Wnk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175579.3 | NP_783169.2 | 4, 5 | 1185 | ||
XM_006247278.2 | XP_006247340.1 | 4, 5 | 1290 | |||
XM_006247279.2 | XP_006247341.1 | 4, 5 | 1290 | |||
XM_006247280.2 | XP_006247342.1 | 5, 6 | 697 | |||
XM_006247281.2 | XP_006247343.1 | 3, 4 | 449 | |||
GenBank mRNA | AY187039.1 | - | 4, 5 | 1162 | ||
AY192567.1 | - | 4, 5 | 1185 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
