siRNA Details
Gene Symbol: Rdm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105842.1 | NP_001099312.1 | 7 | 831 | ||
GenBank mRNA | FQ215272.1 | - | 7 | 764 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rdm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105842.1 | NP_001099312.1 | 7 | 831 | ||
GenBank mRNA | FQ215272.1 | - | 7 | 764 |